2-Amino-3-hydroxybenzoic acid ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-Hydroxyanthranilic acid
A0316-1G
A0316-5G
TCIAA0316-1GEA
90
EUR
TCIAA0316-1G
TCIAA0316-5G
2-Amino-3-hydroxybenzoic acid ≥98.0% (by HPLC, titration analysis)
2-Amino-3-hydroxybenzoic acid
Formula:
C₇H₇NO₃ Peso molecolare: 153,14 g/mol Punto di ebollizione: 357 °C (1013 hPa) Punto di fusione: 240°C |
MDL Number:
MFCD00007700 CAS Number: 548-93-6 EINECS: 208-962-5 |
Risultati del test di specificazione
Appearance | White to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in dilute HCl | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...