Stampa…

3-Bromo-5-fluorobenzonitrile ≥98%

Fornitore: Thermo Scientific
Pericolo

H26066.09 H26066.03
H26066.09EA 144 EUR
H26066.09 H26066.03
3-Bromo-5-fluorobenzonitrile ≥98%
3-Bromo-5-fluorobenzonitrile

Purity: 98% CAS No.: 179898-34-1 Molecular Formula: C7H3BrFN Formula Weight: 200.01 UN No.: 3439 MDL No.: MFCD04038227

Formula: C₇H₃BrFN
Peso molecolare: 200,01 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD04038227
CAS Number: 179898-34-1
UN: 3439
ADR: 6.1,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR