Stampa…
Amantadine hydrochloride 99+%
Fornitore: Thermo Scientific
Attenzione
Synonyms: 1-Adamantylamine hydrochloride, 1-Aminotricyclo[3.3.1.1(3,7)]decane hydrochloride, 1-Aminotricyclo(3.3.1.1(3,7))decan HCl, 1-Aminoadamantane hydrochloride, 1-Adamantanammonium chloride, 1-Adamantanamine hydrochloride, Tricyclo[3.3.1.1(3,7)]dec-1-ylamine hydrochloride, Adamantan-1-amine hydrochloride, 1-Adamantaneammonium Chloride
151670250 151671000
ACRO151670250EA 57.6 EUR
ACRO151670250 ACRO151671000
Amantadine hydrochloride 99+%
Amantadine hydrochloride
Formula: C₁₀H₁₈ClN
Peso molecolare: 187,71 g/mol
Punto di fusione: >300 °C
MDL Number: MFCD00074723
CAS Number: 665-66-7
EINECS: 211-560-2
Visualizza i codici
Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥99.0 %
HPLC ≥96.0 %