Aldeide valerianica ≥97%
Fornitore: Thermo Scientific
|
Pericolo
|
Synonyms:
Pentylaldehyde, Amylaldehyde, Pentanal
Purity: 97%
CAS No.: 110-62-3
Molecular Formula: C5H10O
Formula Weight: 86.13
Flash Point: 8°(46°F)
Refractive Index: 1.3950
Odour: Like aldehyde
Melting Point: -92°
Boiling Point: 102-103°
Beilstein Reference: 1616304
Merck Reference: 14,9902
UN No.: 2058
MDL No.: MFCD00007026
Formula:
C₅H₁₀O Peso molecolare: 86,13 g/mol Punto di ebollizione: 102…103 °C Punto di fusione: –92 °C Densità: 0,810 Flash Pt: 8 °C (46 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00007026 CAS Number: 110-62-3 EINECS: 203-784-4 UN: 2058 ADR: 3,II Merck Index: 14,9902 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...