Tin(II) chloride hydrate ≥99.995% (metals basis), Puratronic®
Fornitore: Thermo Fisher Scientific
|
Pericolo
|
Synonyms:
Tin dichloride hydrate, Stannous chloride hydrate
Tin(II) chloride is widely used as a reducing agent in organic synthesis, for example conversion of quinones into hydroquinones, nitroaromatics into aromatic amines, and in Stephen reduction.
It can be used to reduce certain metal salts into metals, tin plating, analytical testing of mercury ions, formation of silver mirrors and metal-metal bonds. It is also used for tin-plating, as a mordant in textile dyeing and as a catalyst in the production of polylactic acid. Further it is used as a food additive, wherein it serves as an anti-oxidant and a color retention agent. It can be used in the condensation of aryl aldehydes with cyclohexan-1,3-diones to synthesize xanthenes. It catalyzes direct conversion of aldehydes into beta-keto esters.
Formula:
SnCl₂·2H₂O Punto di ebollizione: 623 °C (1013 hPa) Punto di fusione: 43…46 °C Densità: 2,71 Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00149863 CAS Number: 10025-69-1 EINECS: 231-868-0 UN: 3260 ADR: 8,III Merck Index: 14,8783 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...