Stampa…
Bottiglie, a bocca larga, con tappo a vite, Nalgene®
Fornitore: Thermo Fisher Scientific
312105-0016 312105-0004 312105-0008 312105-0001 312105-0032
215-2792EA 677 EUR
215-2792 215-2773 215-2774 215-2771 215-2793
Bottiglie, a bocca larga, con tappo a vite, Nalgene®
Bottiglie Bottiglie in plastica
PPCO, translucent with PP screw cap.
  • Very good chemical resistance
  • Heat resistant up to +121 °C, autoclavable
  • Leakproof
Can be used anywhere in the laboratory and for sample collection. Wide opening for rapid filling and dispensing of powders, pastes and liquids.