Stampa…
Quality monitors, 37 mm
Fornitore: Cytiva (Formerly Pall Lab)
4718 4717
516-0425EA 420 EUR
516-0425 516-0424
Quality monitors, 37 mm
Filtri Filtri a cassetta
Compact and disposable filtration units for analysis of process fluids.
  • Monitor high purity water systems for microbial contamination
Membrane: GN-6 Metricel® (mixed cellulose esters), Metricel® Black (modified polyethersulfone)
Filtration area: 9,1 cm²
Operating temperature: 20 to 25 °C