Stampa…
Storage racks for cryoboxes
Fornitore: GLW
E204 E205 E232 E242 E505 E506 E507 E508 E509 E510 E511 E512 E513 E533 E534 E535 E544 E545 E705 E706 E707 E708 E709 E733 E743 E904 E905 E906 E907 E932 E942 E514 E525 E524
479-1575EA 91.4 EUR
479-1575 479-1576 479-1577 479-1578 479-1579 GLWAE506 479-1580 479-1581 479-1582 479-1583 479-1584 479-1585 479-1586 479-1587 479-1588 479-1589 479-1590 479-1591 479-1592 479-1593 479-1594 479-1595 479-1596 479-1597 479-1598 479-1599 479-1600 479-1601 479-1602 479-1603 479-1604 GLWAE514 GLWAE525 GLWAE524
Storage racks for cryoboxes
Rack Supporti per cryobox
Stainless steel.
  • For 130×130 mm cryoboxes
  • With handle
Fit any freezer. For easy transport and freezing of samples down to −90 °C.