Potassium 98%, chunks in mineral oil
Fornitore: Thermo Scientific
|
Pericolo
|
Purity: 98%
CAS No.: 7440-09-7
Molecular Formula: K
Formula Weight: 39.10
Physical Form: Solid
Density: 0.860
Appearance: Silver/grey
Odour: Odourless
Melting Point: 63-64°
Boiling Point: 765-770°
Merck Reference: 14,7604
UN No.: 2257
MDL No.: MFCD00133776
Formula:
K Peso molecolare: 39,1 g/mol Punto di ebollizione: 765…770 °C Punto di fusione: 63…64 °C Densità: 0,860 Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00133776 CAS Number: 7440-09-7 EINECS: 231-119-8 UN: 2257 ADR: 4.3,I Merck Index: 14,7604 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...