Stampa…
Piastre filtranti, 1,0 ml, per la purificazione dei plasmidi, Nunc™
Fornitore: Thermo Fisher Scientific
278011 278012
NUNC278011EA 1124 EUR
NUNC278011 NUNC278012
Piastre filtranti, 1,0 ml, per la purificazione dei plasmidi, Nunc™
Micropiastre Piastre filtranti
Designed specifically for high throughput sample processing, these filter plates are suitable for vacuum filtration, centrifugation or positive pressure techniques.
  • Shared wall configuration gives reduced plate height, wider wells for faster pocessing, higher yields and less clogging
  • Polyethylene terapthalate (PET) frit in fritted plate has 20 μm pore size
  • Glass fibre plate has PET frit and two layers of glass fibre membrane for binding DNA
  • Purified DNA is suitable for downstream applications, such as sequencing and restriction digestion