Stampa…
Piastre multipozzetto con poli-L-lisina e laminina Corning® BioCoat™
Fornitore: Corning
354595 354596 354619
734-0229EA 284 EUR
734-0229 734-0230 734-0236
Piastre multipozzetto con poli-L-lisina e laminina Corning® BioCoat™
Micropiastre Piastre per colture cellulari
PS, coated with mouse laminin. Laminin, a major structural component of basement membranes, has many varied functions that are mediated by binding to various components of the basement membrane (for example, collagen IV) and to cell-surface receptors. Laminin-coated cell culture plates have a number of applications, including the promotion of cell adhesion, proliferation and differentiation of a variety of cell types, particularly neurons, epithelial cells, myocytes and myoblasts.
Laminin source: Engelbreth-Holm-Swarm (EHS) mouse tumor. Tested for ability to initiate differentiation (More Product Information
Visualizza i codici
Specifiche
Environmentally Preferable Produzione a basso impatto
Certificazione ambientale ISO 14001 - Bedford