Stampa…

(-)-Chinina ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: (R)-[(2S,4S,5R)-1-Aza-5-vinylbicyclo[2.2.2]oct-2-yl](6-methoxyquinolin-4-yl)methanol, Quinine, (R)-(6-methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol

Q0028-25G Q0028-100G
TCIAQ0028-25GEA 56 EUR
TCIAQ0028-25G TCIAQ0028-100G
(-)-Chinina ≥98.0% (by titrimetric analysis)
(-)-Chinina
Formula: C₂₀H₂₄N₂O₂
Peso molecolare: 324,42 g/mol
Punto di fusione: 176 °C
MDL Number: MFCD00198096
CAS Number: 130-95-0
EINECS: 205-003-2
Merck Index: 14,08061
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1862e14275fd2e1c20e8b391d12ec26627315de8&vid=1862e14275fd2e1c20e8b391d12ec26627315de8&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 174.0 to 179.0 deg-C
Specific rotation [a]20/D -160 to -170 deg (C=2, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR