Stampa…
Lavavetreria termodisinfettante, G 7883 CD
Fornitore: MIELE
5637290 7565680 5806990 5807110 5764970 5774550 5764890 5799650 5774680 5798640
141-0948EA 0 EUR
141-0948 141-0583 141-0465 141-1004 141-0444 141-0449 141-0443 141-0463 141-0452 141-0455
Lavavetreria termodisinfettante, G 7883 CD
Lavavetreria
The G 7883 CD cleaner/disinfector is suitable for washing laboratory glassware for analytical experiments in eg industry, environmental and reseach laboratories. This appliance has a large two-level wash cabinet and can be integrated into a continuous work top as a built-under unit or installed as a freestanding unit. The G 7883 CD differs from the G 7883 in that it has an active drying unit which ensures drying inside and outside of the items washed via a hot air blower. In addition, the G 7883 CD is 90 cm wide and has a front-loading drawer for 2 reservoirs.
  • Direct connection of the upper basket and mobile units to the hydraulic circuit enables thorough cleaning inside and outside of the items washed
  • Integrated dispensing pump for precise and automatic dispensing of liquid agents
  • Final rinse with demineralised water for results suitable for analytical experiments
  • Profi-Monobloc ensures continual regeneration during cleaning
  • Steam condenser ensures that the room is not exposed to steam and hot air
  • Unnecessary to connect an exhaust duct
Con unità di asciugatura dinamica.
In accordance with VDE, EMC/radio interference protection, DVGW, IP X1.
Certificazioni: Conforme alla norma VDE, EMC/protezione dalle interferenze radio, DVGW, IP X1.
Informazioni per l'ordine: i cestelli devono essere ordinati separatamente. Altri modelli disponibili su richiesta, ad esempio con rivestimento bianco o con bordo decorativo. Per uso come unità indipendente, ordinare separatamente la copertura.