Stampa…
FT-IR purge gas generators
Fornitore: PARKER HANNIFIN
75-62UK 75-45UK 75-52UK
559-0012EA 7030 EUR
559-0012 PARK75-45UK PARK75-52UK
FT-IR purge gas generators
Generatori di gas Generatori di anidride carbonica
These purge gas generators are specifically designed for use with FT-IR Spectrometers to provide a purified purge gas and air bearing gas from compressed air. The generators supply carbon dioxide-free air at less than −70 °C dew point with no suspended impurities larger than 0,1 µm. They are also ideally suited for use with CO₂ -analysers and Matrix GCs in addition to supplying other laboratory instruments.
  • Eliminate the need for costly, inconvenient nitrogen cylinders in the laboratory
  • Constant purge at high flow rate, 24 hours/day, 7 days/week
  • Protect beam splitter from corrosion by moisture
  • No delay in operation while instrument is being purged
The generators are quiet, reliable and easy to install - simply attach the inlet and outlet air lines, plug the power cord into a wall outlet and enjoy trouble-free operation.
CO₂ concentration: <1 ppm.
Inlet/outlet port size: ¼" NPT female.
Max. inlet temperature: 25 °C.
Min./max. inlet air pressure: 4/8,5 bar.