Stampa…
Portable autoclaves, MLS Series, PHCbi
Fornitore: PHC Corporation
MLS-3020U MLS-3751L
481-0507EA 0 EUR
481-0507 481-0687
Portable autoclaves, MLS Series, PHCbi
Sterilizzatori Autoclavi (sterilizzatori a vapore)
Energy saving top loading autoclaves provide a safe, reliable high pressure steam sterilising environment within a self contained unit. Safety assured with audible and visual alarm alerts, circuit breaker, pressure safety valve and fail safe functions such as overheating protection, door/lid interlock and scorch prevention. Microprocessor controls ensure that the temperature is accurately maintained.
  • Available in four sizes, 12 pre-set programmes for sterilising, preparing and maintaining culture media (MLS-3750/3780)
  • Portable with wheels, compact design