Stampa…
Piastre di protezione da laboratorio in vetroceramica, DURAN®
Fornitore: DWK Life Sciences
238215309 238215703 238215806
451-9421EA 36.5 EUR
451-9421 451-9422 451-9423
Piastre di protezione da laboratorio in vetroceramica, DURAN®
Vassoi/Piatti/Contenitori Vassoi/piatti per laboratorio
These quadrupods are made from heat-resistant chrome-nickel steel with four legs for extra stability.
  • Uniformity of heating distribution
  • Excellent energy transfer
  • Plates can either be used with quadrupods or with plate holders
Temperature: −200 to +700 °C