Stampa…
Syringes, medical
Fornitore: CODAN
627704 628426 623760 624717 626706 622611
CODA627704EA 87.9 EUR
CODA627704 613-4236 613-4231 613-4232 613-4234 613-0098
Syringes, medical
Siringhe Siringhe, uso medico
Sterile, graduated, single use, three pieces, body made of transparent PP, white plunger in PP/titanium oxide and with silicone seal, no latex.
  • Standard Luer Tip (LT) or screw connector Luer Lock (LL), centred (1 to 3 ml) or off-centred (5 to 100 ml)
  • The watertight silicone seal reduces contact with the solution and guarantees minimal risk of particulate contamination of the solution
  • Sterilised with ethylene oxide (EN 550)
Certificazioni: Conforms to ISO 9001, EN 46001, medical guideline 93/42/EEC
Confezione: In individual blister packs.