Stampa…
Electronic diamond scriber, EDS 20
Fornitore: MEDITE GMBH
46-4030-00
811-0223EA 0 EUR
811-0223
Electronic diamond scriber, EDS 20
Penne Penne stilo e punte
For inscribing object holders and marking, inscribing or etching metal or plastic. Manually bringing the scribe into the writing position will automatically activate the motor; when the scribe is returned to a horizontal position, the motor switches itself off. When in the writing position, the tip rotates very quickly, thereby enabling effortless etching. Weight approx. 100 g, average finger grips approx. 15 mm.
  • Tips are light and can be replaced without the need for tools
  • Inscription is etched so deep that it can last almost indefinitely