Stampa…
Spoon spatulas
Fornitore: BOCHEM
3210 3211 3212 3213 3214 3215
231-1050EA 4.8 EUR
231-1050 231-1325 231-1328 231-1051 231-1052 231-1053
Spoon spatulas
Spatole
Stainless steel. Universal spoon spatula designed for a wide range of uses.
  • One end is shaped like a spoon and the other end like a spatula
  • With rounded handle
  • High thermal resistance, autoclavable
  • Excellent resistance to chemicals
  • Quick and easy to clean