Stampa…
Signs, fire signs
Fornitore: KROSCHKE
Z5887 W17027 W17033 W2395 Z5908 Z5905
116-1140EA 203 EUR
116-1140 116-2016 116-2019 116-2024 116-2026 116-2027
Signs, fire signs
Segnali
These self-adhesive, PVC film signs are used to indicate fire equipment. According to EC Directive 92/58/EEC and related national legislation on safety and/or health signs and also according to to VBG 125 (a regulation of the German employer's liability insurance association), signboards/signs for fire equipment must be permanently fitted and clearly recognisable at all times.
  • Suitable for use inside and outside
  • Visible from a distance of ∼12 metres
  • Long-lasting persistent luminescence material available
Certificazioni: VBG 125 (now known as BGV A8).
Attenzione: Long-lasting persistent luminescence fire signs must be displayed in new and refurbished buildings if no safety lighting is available.