Stampa…
Scalpel handles
Fornitore: BAYHA
FIGUR1 FIGUR4 FIGUR5
233-5201EA 47.1 EUR
233-5201 233-5202 233-5203
Scalpel handles
Bisturi Manici per bisturi
Acciaio inox.
  • Available in solid form or as version with hollow handle (233-5203)
  • Large selection of sterile and non sterile scalpel blades available; all blade shapes fit into any BAYHA handle
  • Proven latch system for tight fit of the blade in the handle
  • Easy and safe exchange of scalpel blades