Stampa…
Pipette graduate, coltura tessuti
Fornitore: HIRSCHMANN
1170135 1170142 1170154 1170160 1170171
612-1087EA 8.1 EUR
612-1087 612-1088 612-1089 612-1090 612-1099
Pipette graduate, coltura tessuti
Pipette Pipette graduate
AR-Glas®.
  • Calibrated to deliver 'Ex' (TD)
  • Marks and inscriptions in durable amber stain
  • With standard cotton plug end (OD 8 mm)
  • Ideal for tissue culture and working on cleanroom benches
Lunghezza: 230 mm ±3 mm.