Stampa…
Piastre PCR per qPCR, 24 e 38 pozzetti
Fornitore: Brand
781411 781415 781412 781416
731-0014EA 83.7 EUR
731-0014 731-0015 731-0165 731-0166
Piastre PCR per qPCR, 24 e 38 pozzetti
Micropiastre Piastre per PCR
PP, non skirted. Manufactured under cleanroom conditions, and made from pure, high quality materials (without additives) these PCR plates are suitable for use with most commercially available thermal cyclers.
  • Ultra-thin walled design to facilitate constant, rapid and precise heat transfer
  • Smooth vessel interior minimises the binding of enzymes and nucleic acid to the walls
  • Alphanumeric coding embossed and stamped in blue
  • Raised well rims protect against cross-contamination and provide a tight seal in combination with matching sealing mat
  • Free from DNase, DNA and RNase
  • Autoclavable at 121 °C (2 bar), acc. DIN EN 285