Stampa…
Industrial gloves, Nitrotough™ N630
Fornitore: Marigold (Ansell)
200PB0508
MARG200PB0508EA 0 EUR
MARG200PB0508
Industrial gloves, Nitrotough™ N630
Guanti
Heavy duty palm coated nitrile gloves are suitable for use in automotive, building and fabrication. Reusable and washable.
  • High cut and abrasion resistance
  • Excellent dry grip properties
  • 100% cotton fleeced liner
  • Open backed for increased comfort when working in warm conditions
Typical applications include: Press shop operations, handling structural and body components, handling materials with sharp or rough edges.
Certificazioni: EN 388:2016 (4221B); Cat. II