Stampa…
Filtration Crucibles
Fornitore: ROBU GLASFILTER
20082 20083 20084 20151 20152 20153 20154 20301 20302 20304 20305 20501 20502 20503 20504 20505 20155 20080 20081 20150 20300 20500 20085 2050C 2030C 2030F 20303 2050F
511-2102EA 15.9 EUR
511-2102 511-2103 511-2104 511-2201 511-2202 511-2203 511-2204 511-2301 511-2302 511-2304 511-2305 511-2401 511-2402 511-2403 511-2404 511-2405 511-0360 511-0358 511-2101 511-0359 511-0249 511-0361 511-0248 ROBU2050C ROBU2030C ROBU2030F 511-0273 ROBU2050F
Filtration Crucibles
Crogioli
ROBU® Filter Crucibles with intergrated VitraPOR® filter provide efficient filtration for laboratory applications. Made from durable materials, these crucibles are designed for high-performance filtration, offering reliable separation of solids and liquids. Ideal for gravimetric analysis and other precise filtration processes.
  • Durable and resistant to high temperatures
  • Efficient filtration
  • Ideal for gravimetric analysis
  • Ensures precise separation of solids and liquids
  • Available in various sizes
  • Compatible with most filtration setups
  • Marked with an individual number for distinct sample identification
  • Material: Borosilicate glass 3.3
ROBU Filter Crucibles with intergrated VitraPOR filter are engineered for efficient and precise filtration in laboratory environments. Made from high-quality, durable materials, these crucibles offer exceptional resistance to high temperatures and harsh chemicals, ensuring reliability and longevity even in demanding filtration processes. They are perfect for use in gravimetric analysis and other applications that require the separation of solids from liquids. With easy-to-clean and reusable features, these crucibles provide long-term value and consistent performance. Available in various sizes, they are designed to fit most filtration setups, making them a versatile and essential tool in laboratories worldwide.
Certificazioni: In compliance with ISO 4796-80 and ASTM E128-99 standards.