Stampa…
Tubing connectors, mini, Y-shape
Fornitore: PIEPER FILTER
PP-S16S16Y PP-S24S24Y PP-S32S32Y
229-0501EA 67.7 EUR
229-0501 229-0502 229-0503
Tubing connectors, mini, Y-shape
Raccordi e connettori
PP, translucent.
  • Very good thermal resistance
  • Very good chemical resistance
  • Robust, durable
Perfect for conneting tubing with small inner diameters