Stampa…
Spare screw caps for bottles with wide mouth
Fornitore: KJS KUNSTSTOFFVERPACKUNGEN
V40E V50E V65E
215-6024EA 2.2 EUR
215-6024 215-6025 215-6026
Spare screw caps for bottles with wide mouth
Ghiere Tappi per bottiglie
LDPE, natural.
  • Good chemical resistance
  • Good thermal resistance
  • Leakproof
Screw caps are only suitable for bottles 215-5630 to 215-5636.