Stampa…
Bottiglie da laboratorio, collo stretto, con tappi a vite, PYREX®
Fornitore: SciLabware
1515/08D 1515/10D 1515/12D 1515/16D 1515/18D 1515/01D 1515/04D 1515/06D 1515/22D
215-0334EA 17.2 EUR
215-0334 215-0335 215-0336 215-0337 215-0338 215-0330 215-0332 215-0333 BIBB1515/22D
Bottiglie da laboratorio, collo stretto, con tappi a vite, PYREX®
Bottiglie Bottiglie in vetro
PYREX®, borosilicate glass with phenolic screw caps.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Round, with white graduations and marking spot. Ideal for storage of reagents, analytical standards and culture media etc. Supplied with phenolic screw caps, having a PTFE-faced sealing disc. With printed retrace code (batch identification); batch certificate available at www.scilabware.com.
* Not graduated
Certificazioni: ISO 4796