Stampa…
Pellicola per chemiluminescenza, Kodak BioMax® Light
Fornitore: CARESTREAM HEALTH
1538966 1917012 8689358 8761520
730-3211EA 0 EUR
730-3211 730-3226 730-3238 730-3244
Pellicola per chemiluminescenza, Kodak BioMax® Light
Pellicole per autoradiografie Pellicole
BioMax® Light is a double emulsion film that provides maximum clarity and sensitivity for detection of chemiluminescent-labelled samples. BioMax® Light produces the highest signal-to-noise ratio of any Kodak film for detection of chemiluminescence. This film performs well with all brands of chemiluminescent kits. BioMax® Light film is suitable for direct and intensifying screen exposures with commonly used isotopes, though it provides lower sensitivity resulting in longer exposure times compared to BioMax® MS, and X-OMAT AR films for radioisotope detection.
  • Suitable for both manual and