Stampa…
VWR®, connettori a soffietto per giunti di terra standard
Fornitore: VWR Collection
201-0062EA 52.5 EUR
201-0062 201-0063 201-0064 201-0065 201-0066 201-0067 201-0068
VWR®, connettori a soffietto per giunti di terra standard
Adattatori Adattatori conici a smerigliatura
In PTFE, con giunti smerigliati sul cono e sul manicotto.
  • Eccellente resistenza chimica
  • A tenuta di gas, liquidi e vuoto
  • Resistente a temperature da −200 a +280 °C
For use as a flexible or elastic connector to prevent pressure and stresses, and to dampen vibrations in standard ground joints. Sturdy construction with sealing rings, which prevent the ground joint from seizing and give a very good seal. No grease required, so joint grease residue cannot contaminate the analysis. Can be bent to an angle of 120°.