L-Phenylalaninol ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
(S)-(-)-2-Amino-3-phenyl-1-propanol, (S)-(-)-Phenylalaninol, (S)-2-Amino-3-phenylpropan-1-ol, L(-)-2-Amino-3-phenyl-1-propanol, L(-)-Phenylalaninol, H-Phenylalaninol, L(-)-2-Amino-3-phenylpropan-1-ol, (S)-(-)-2-Amino-3-phenylpropan-1-ol
P1028-25G
P1028-5G
TCIAP1028-25GEA
131
EUR
TCIAP1028-25G
TCIAP1028-5G
L-Phenylalaninol ≥98.0% (by GC, titration analysis)
L-Phenylalaninol
Formula:
C₉H₁₃NO Peso molecolare: 151,21 g/mol Punto di ebollizione: 278 °C (1013 hPa) Punto di fusione: 91…94 °C |
MDL Number:
MFCD00004732 CAS Number: 3182-95-4 EINECS: 221-674-4 UN: 3259 ADR: 8,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 91.0 to 95.0 deg-C |
Specific rotation [a]20/D | -24.0 to -25.0 deg (C=5, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...