Stampa…

Esomeprazole magnesium trihydrate 98%

Fornitore: Thermo Fisher Scientific
Attenzione

472510010 472512500
472510010.EA 401 EUR
472510010. 472512500.
Esomeprazole magnesium trihydrate 98%
Esomeprazole magnesium trihydrate
Formula: C₃₄H₄₂MgN₆O₉S₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD31807353
CAS Number: 217087-09-7
UN: 3077
ADR: 9,III

Visualizza i codici

Risultati del test di specificazione

Appearance white to light yellow powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Optical rotation -128° to -146° (c=0.5,Methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR