Stampa…

Ledipasvir acetone 98%

Fornitore: Thermo Fisher Scientific

469361000 469365000
469361000.EA 72.7 EUR
469361000. 469365000.
Ledipasvir acetone 98%
Ledipasvir acetone
Formula: C₄₉H₅₄F₂N₈O₆·C₃H₆O
MDL Number: MFCD28167732
CAS Number: 1441674-54-9

Visualizza i codici

Risultati del test di specificazione

Appearance white to yellow powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR