Costunolide
Fornitore: Thermo Fisher Scientific
Synonyms:
(3aS,6E,10E,11aR)-6,10-Dimethyl-3-methylene-3a,4,5,8,9,11a-hexahydrocyclodeca[b]furan-2(3H)-one
Formula:
C₁₅H₂₀O₂ Peso molecolare: 232.32 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00210262 CAS Number: 553-21-9 |
Risultati del test di specificazione
Appearance | white to almost white powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Melting point | 104°C to 109°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...