Palmatine chloride 97%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
467150010
467152500
467150010.EA
308
EUR
467150010.
467152500.
Palmatine chloride 97%
Palmatine chloride
Formula:
C₂₁H₂₂ClNO₄ Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00016656 CAS Number: 10605-02-4 |
Risultati del test di specificazione
Appearance | light yellow to yellow to orange crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Loss on drying | ≤5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...