Stampa…

Dinotefuran

Fornitore: Thermo Fisher Scientific
Attenzione

467090010 467092500
467090010.EA 117 EUR
467090010. 467092500.
Dinotefuran
Dinotefuran
Formula: C₇H₁₄N₄O₃
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD06795001
CAS Number: 165252-70-0
UN: 3077
ADR: 9,III

Visualizza i codici

Risultati del test di specificazione

Appearance white to almost white powder or crystals
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 100°C to 108°C
Water ≤1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR