Stampa…
Vanadile solfato pentaidrato 17-23% (v/v)
Fornitore: Thermo Fisher Scientific
Attenzione
Synonyms: Basic vanadium (IV) sulphate pentahydrate, Vanadium oxide sulphate pentahydrate, Vanadium (IV) sulphate oxide pentahydrate
214841000 214840250
ACRO214841000EA 208 EUR
ACRO214841000 ACRO214840250
Vanadile solfato pentaidrato 17-23% (v/v)
Vanadile solfato pentaidrato
Formula: VOSO₄·5H₂O
Peso molecolare: 253.08 g/mol
Densità: 2
MDL Number: MFCD00149933
CAS Number: 12439-96-2
EINECS: 248-652-7
UN: 2931
ADR: 6.1 II
Visualizza i codici
Appearance Blue Moist crystalline powder or crystals
Degree of hydration ca. 5 moles water
Titration with KMnO4 17 to 23 % (V)
Iron (Fe) ≤600 ppm
Potassium (K) ≤500 ppm
Sodium (Na) ≤500 ppm