Stampa…

Gel di silice, Pore size: 60 Å, LiChroprep® for liquid chromatography

Fornitore: MERCK PRODUCTION CHEMICALS

1.09390.0100 1.09390.9025 1.09390.9030
1.09390.0100EA 0 EUR
1.09390.0100 1.09390.9025 1.09390.9030
Gel di silice, Pore size: 60 Å, LiChroprep® for liquid chromatography
Gel di silice
Formula: SiO₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011232
CAS Number: 63231-67-4

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR