Stampa…
X-Gal (5-Bromo-4-cloro-3-indolil-ß-D-galattopiranoside) 99+% Molecular_Biology DNAse_RNAse_PROTEase_free
Fornitore: Thermo Fisher Scientific
Synonyms: BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
327241000 327245000
ACRO327241000EA 65.3 EUR
ACRO327241000 ACRO327245000
X-Gal (5-Bromo-4-cloro-3-indolil-ß-D-galattopiranoside) 99+% Molecular_Biology DNAse_RNAse_PROTEase_free
X-Gal (5-Bromo-4-cloro-3-indolil-ß-D-galattopiranoside)
Formula: C₁₄H₁₅BrClNO₆
Peso molecolare: 408.63 g/mol
Punto di fusione: 230 °C (dec.)
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00005666
CAS Number: 7240-90-6
EINECS: 230-640-8
Visualizza i codici
Appearance White Powder
Infrared spectrum Conforms
Water ≤0.2 %
TLC ≥99.0 %
Specific optical rotation -61° to -63° (20°C, 589 nm) (c=1, H2O/DMF 1/1)
Dnase,Rnase,Protease activity none detected