Stampa…

1,3-Thiazolan-2-one

Fornitore: TCI

T3926-25G T3926-5G
TCIAT3926-25GEA 89 EUR
TCIAT3926-25G TCIAT3926-5G
1,3-Thiazolan-2-one
1,3-Thiazolan-2-one
Formula: C₃H₅NOS
MDL Number: MFCD01332388
CAS Number: 2682-49-7

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR