Stampa…

4,4',4''-(Benzene-1,3,5-triyltris(oxy))trianiline

Fornitore: TCI EUROPE

Synonyms: 1,3,5-Tris(4-aminophenoxy)benzene

T3909-1G T3909-200MG
TCIAT3909-1GEA 228 EUR
TCIAT3909-1G TCIAT3909-200MG
4,4',4''-(Benzene-1,3,5-triyltris(oxy))trianiline
4,4',4''-(Benzene-1,3,5-triyltris(oxy))trianiline
Formula: C₂₄H₂₁N₃O₃
Peso molecolare: 399,44 g/mol
MDL Number: MFCD09908237
CAS Number: 102852-92-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR