Thiamethoxam
Fornitore: TCI
T3811-1G
T3811-25G
T3811-5G
TCIAT3811-1GEA
68
EUR
TCIAT3811-1G
TCIAT3811-25G
TCIAT3811-5G
Thiamethoxam
Thiamethoxam
Formula:
C₈H₁₀ClN₅O₃S |
MDL Number:
MFCD03792862 CAS Number: 153719-23-4 UN: 1325 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...