Stampa…

Pimobendan

Fornitore: TCI

P2779-250MG P2779-50MG
TCIAP2779-250MGEA 148 EUR
TCIAP2779-250MG TCIAP2779-50MG
Pimobendan
Pimobendan
Formula: C₁₉H₁₈N₄O₂
MDL Number: MFCD00761648
CAS Number: 74150-27-9

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR