Stampa…

L-β-Homoproline hydrochloride

Fornitore: TCI

P2777-1G P2777-250MG
TCIAP2777-1GEA 217 EUR
TCIAP2777-1G TCIAP2777-250MG
L-β-Homoproline hydrochloride
L-beta-Homoproline hydrochloride
Formula: C₆H₁₂ClNO₂
MDL Number: MFCD07363484
CAS Number: 53912-85-9

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR