Stampa…

9-Phenyl-3-(2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-9H-carbazole

Fornitore: TCI EUROPE

Synonyms: 9-Phenyl-3-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-9H-carbazole

P2772-1G P2772-5G
TCIAP2772-1GEA 54 EUR
TCIAP2772-1G TCIAP2772-5G
9-Phenyl-3-(2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-9H-carbazole
9-Phenyl-3-(2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-9H-carbazole
Formula: C₃₀H₂₈BNO₂
Peso molecolare: 445,3598 g/mol
MDL Number: MFCD32670129
CAS Number: 1628817-49-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR