Stampa…

4-Amino-5-chloro-N-(1-(3-methoxypropyl)piperidin-4-yl)-2,3-dihydrobenzofuran-7-carboxamide

Fornitore: TCI EUROPE

Synonyms: Prucalopride

P2467-10MG P2467-50MG
TCIAP2467-10MGEA 34 EUR
TCIAP2467-10MG TCIAP2467-50MG
4-Amino-5-chloro-N-(1-(3-methoxypropyl)piperidin-4-yl)-2,3-dihydrobenzofuran-7-carboxamide
4-Amino-5-chloro-N-(1-(3-methoxypropyl)piperidin-4-yl)-2,3-dihydrobenzofuran-7-carboxamide
Formula: C₁₈H₂₆ClN₃O₃
Peso molecolare: 367,87 g/mol
MDL Number: MFCD09837787
CAS Number: 179474-81-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR