Nitrendipine
Fornitore: TCI
Formula:
C₁₈H₂₀N₂O₆ |
MDL Number:
MFCD00082255 CAS Number: 39562-70-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...