3-Nitrophenyl isothiocyanate
Fornitore: TCI
Formula:
C₇H₄N₂O₂S |
MDL Number:
MFCD00041245 CAS Number: 3529-82-6 UN: 2811 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...