Stampa…

4-Nitrophenyl phenyl ether

Fornitore: TCI

Synonyms: 4-Nitrodiphenyl ether, 1-Nitro-4-phenoxybenzene

N1011-25G N1011-5G
TCIAN1011-25GEA 126 EUR
TCIAN1011-25G TCIAN1011-5G
4-Nitrophenyl phenyl ether
4-Nitrophenyl phenyl ether
Formula: C₁₂H₉NO₃
MDL Number: MFCD00007320
CAS Number: 620-88-2
UN: 3077

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR