Stampa…

2-Methyl-2-phenylpropanenitrile

Fornitore: TCI

M3308-1G M3308-5G
TCIAM3308-1GEA 47 EUR
TCIAM3308-1G TCIAM3308-5G
2-Methyl-2-phenylpropanenitrile
2-Methyl-2-phenylpropanenitrile
Formula: C₁₀H₁₁N
MDL Number: MFCD00099614
CAS Number: 1195-98-8
UN: 3276

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR